ADVH14DBDA7F AChemBlock

5-chloro-3-iodo-1-(oxan-2-yl)-1H-pyrazolo[4,3-b]pyridine

  • MDL number MFCD22689818
  • Cas Number : 1416713-15-9
  • Molecular Weight : 363.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-iodo-1-oxan-2-yl-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48330
Availability details for
Quantity
  • Availability
  • Add to Cart