ADVH14DBDCD0 AChemBlock

6-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid

  • MDL number MFCD09880165
  • Cas Number : 872355-55-0
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-1h-pyrrolo-2-3-b-pyridine-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45012
Availability details for
Quantity
  • Availability
  • Add to Cart