ADVH14DBDCD8 AChemBlock

tert-butyl 3,3-difluoro-2-(hydroxymethyl)azetidine-1-carboxylate

  • MDL number MFCD27987674
  • Cas Number : 1638759-62-2
  • Molecular Weight : 223.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-3-difluoro-2-hydroxymethyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47682
Availability details for
Quantity
  • Availability
  • Add to Cart