ADVH14DBDCEF AChemBlock

1,4-dioxaspiro[4.5]decan-8-amine

  • Cas Number : 97096-16-7
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,4-dioxaspiro[4.5]dec-8-ylamine
Inchi Code 1S/C8H15NO2/c9-7-1-3-8(4-2-7)10-5-6-11-8/h7H,1-6,9H2
Inchi Key KDAFVGCPLFJMHY-UHFFFAOYSA-N

Description

Synonym 1,4-dioxaspiro[4.5]decan-8-amine
Purity 0.97
Country of Origin CN
Product Link https://www.achemblock.com/1-4-dioxaspiro-4-5-decan-8-amine.html

Safety Information

MSDS https://www.achemblock.com/ac_sds?Catalog=P44600
Availability details for
Quantity
  • Availability
  • Add to Cart