ADVH14DBDCF1 AChemBlock

3-fluoro-2-methylazetidine hydrochloride

  • MDL number MFCD28501878
  • Cas Number : 1638771-97-7
  • Molecular Weight : 125.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-2-methylazetidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47451
Availability details for
Quantity
  • Availability
  • Add to Cart