ADVH14DBDD65 AChemBlock

5-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1h-pyrrolo[2,3-b]pyridine

  • MDL number MFCD20923099
  • Cas Number : 1259279-57-6
  • Molecular Weight : 416.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1-tosyl-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44020
Availability details for
Quantity
  • Availability
  • Add to Cart