ADVH14DBDD7E AChemBlock

3-ethynyl-1,1-difluorocyclobutane

  • MDL number MFCD30802459
  • Cas Number : 1698054-36-2
  • Molecular Weight : 116.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-ethynyl-1-1-difluorocyclobutane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47348
Availability details for
Quantity
  • Availability
  • Add to Cart