ADVH14DBDDC2 AChemBlock

5-methoxy-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid

  • MDL number MFCD12962882
  • Cas Number : 1190322-32-7
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-1h-pyrrolo-2-3-b-pyridine-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49464
Availability details for
Quantity
  • Availability
  • Add to Cart