ADVH14DBDDF3 AChemBlock

3-fluoro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD15529168
  • Cas Number : 1016241-78-3
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-1h-pyrrolo-2-3-b-pyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49980
Availability details for
Quantity
  • Availability
  • Add to Cart