ADVH14DBDE2F AChemBlock

1-tert-butyl 3-ethyl 3-(hydroxymethyl)azetidine-1,3-dicarboxylate

  • MDL number MFCD31705470
  • Cas Number : 1078166-51-4
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-3-ethyl-3-hydroxymethyl-azetidine-1-3-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49854
Availability details for
Quantity
  • Availability
  • Add to Cart