ADVH14DBDE89 AChemBlock

3-(benzyloxy)bicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD31537335
  • Cas Number : 2166647-54-5
  • Molecular Weight : 218.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-benzyloxy-bicyclo-1-1-1-pentane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46127
Availability details for
Quantity
  • Availability
  • Add to Cart