ADVH14DBDEB5 AChemBlock

1-methyl-2-oxoazetidine-3-carboxylic acid

  • MDL number MFCD19229327
  • Cas Number : 42599-27-9
  • Molecular Weight : 129.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-2-oxoazetidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45627
Availability details for
Quantity
  • Availability
  • Add to Cart