ADVH14DBDEDF AChemBlock

tert-butyl N-[(1S,2S)-2-hydroxycyclobutyl]carbamate

  • MDL number MFCD30802670
  • Cas Number : 2231664-24-5
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1s-2s-2-hydroxycyclobutyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P50892
Availability details for
Quantity
  • Availability
  • Add to Cart