ADVH14DBDEF1 AChemBlock

tert-butyl (2S,3S)-3-amino-2-methylazetidine-1-carboxylate

  • MDL number MFCD31705524
  • Cas Number : 2068138-10-1
  • Molecular Weight : 186.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-3s-3-amino-2-methylazetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46394
Availability details for
Quantity
  • Availability
  • Add to Cart