ADVH14DBDEFA AChemBlock

3,3-dimethoxycyclobutane-1-carbaldehyde

  • MDL number MFCD24686435
  • Cas Number : 1031334-86-7
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethoxycyclobutane-1-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49940
Availability details for
Quantity
  • Availability
  • Add to Cart