ADVH14DBE011 AChemBlock

exo-3-(boc-aminomethyl)-8-azabicyclo[3.2.1]octane

  • MDL number MFCD22397006
  • Cas Number : 455267-38-6
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-3-boc-aminomethyl-8-azabicyclo-3-2-1-octane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43973
Availability details for
Quantity
  • Availability
  • Add to Cart