ADVH14DBE03E AChemBlock

N-(3-aminocyclobutyl)acetamide

  • MDL number MFCD12082072
  • Cas Number : 1156366-87-8
  • Molecular Weight : 128.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-3-aminocyclobutyl-acetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49715
Availability details for
Quantity
  • Availability
  • Add to Cart