ADVH14DBE046 AChemBlock

2-methylazetidin-3-ol hydrochloride

  • MDL number MFCD22570261
  • Cas Number : 920282-10-6
  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methylazetidin-3-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44851
Availability details for
Quantity
  • Availability
  • Add to Cart