ADVH14DBE047 AChemBlock

3-ethylazetidin-3-amine

  • MDL number MFCD11849054
  • Cas Number : 866432-22-6
  • Molecular Weight : 100.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-ethylazetidin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45038
Availability details for
Quantity
  • Availability
  • Add to Cart