ADVH14DBE06A AChemBlock

tert-butyl 3-{[(benzyloxy)carbonyl]amino}-3-(hydroxymethyl)azetidine-1-carboxylate

  • MDL number MFCD23105885
  • Cas Number : 1363380-94-2
  • Molecular Weight : 336.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-benzyloxy-carbonyl-amino-3-hydroxymethyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43914
Availability details for
Quantity
  • Availability
  • Add to Cart