ADVH14DBE0BB AChemBlock

4-fluoro-1H-pyrrolo[2,3-b]pyridin-5-ol

  • MDL number MFCD11656012
  • Cas Number : 651744-21-7
  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1h-pyrrolo-2-3-b-pyridin-5-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44378
Availability details for
Quantity
  • Availability
  • Add to Cart