ADVH14DBE0EA AChemBlock

6-chloro-4-nitro-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD15529248
  • Cas Number : 892414-50-5
  • Molecular Weight : 265.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-nitro-3-trifluoromethyl-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44939
Availability details for
Quantity
  • Availability
  • Add to Cart