ADVH14DBE0EB AChemBlock

methyl 6-bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylate

  • MDL number MFCD20923471
  • Cas Number : 1342811-51-1
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromo-1h-pyrrolo-2-3-b-pyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48712
Availability details for
Quantity
  • Availability
  • Add to Cart