ADVH14DBE118 AChemBlock

tert-butyl 3-(aminomethyl)-3-{[(tert-butoxy)carbonyl]amino}azetidine-1-carboxylate

  • MDL number MFCD28502471
  • Cas Number : 1936305-61-1
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-aminomethyl-3-tert-butoxy-carbonyl-amino-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46558
Availability details for
Quantity
  • Availability
  • Add to Cart