ADVH14DBE180 AChemBlock

ethyl 1-(fluoromethyl)-4-oxocyclohexane-1-carboxylate

  • MDL number MFCD30802885
  • Cas Number : 1818868-39-1
  • Molecular Weight : 202.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-fluoromethyl-4-oxocyclohexane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46974
Availability details for
Quantity
  • Availability
  • Add to Cart