ADVH14DBE1B1 AChemBlock

exo-tert-butyl-8-hydroxy-3-azabicyclo[3.2.1]octane-3-carboxylate

  • MDL number MFCD30486378
  • Cas Number : 2101733-97-3
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-tert-butyl-8-hydroxy-3-azabicyclo-3-2-1-octane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46246
Availability details for
Quantity
  • Availability
  • Add to Cart