ADVH14DBE1B6 AChemBlock

bicyclo[3.2.1]octane-3-carboxylic acid

  • MDL number MFCD19981843
  • Cas Number : 6221-65-4
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/bicyclo-3-2-1-octane-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45392
Availability details for
Quantity
  • Availability
  • Add to Cart