ADVH14DBE1D9 AChemBlock

tert-butyl N-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-en-1-yl]carbamate

  • MDL number MFCD12032216
  • Cas Number : 1251732-64-5
  • Molecular Weight : 323.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-tetramethyl-1-3-2-dioxaborolan-2-yl-cyclohex-3-en-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44342
Availability details for
Quantity
  • Availability
  • Add to Cart