ADVH14DBE22B AChemBlock

4-bromocyclohexan-1-ol

  • MDL number MFCD20624182
  • Cas Number : 89599-47-3
  • Molecular Weight : 179.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromocyclohexan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44927
Availability details for
Quantity
  • Availability
  • Add to Cart