ADVH14DBE23F AChemBlock

(1r,4r)-4-hydroxy-1-methylcyclohexane-1-carboxylic acid

  • MDL number MFCD30803003
  • Cas Number : 76657-91-5
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-4-hydroxy-1-methylcyclohexane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45202
Availability details for
Quantity
  • Availability
  • Add to Cart