ADVH14DBE240 AChemBlock

4-(aminomethyl)cyclohexan-1-ol

  • MDL number MFCD09745276
  • Cas Number : 164646-07-5
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-cyclohexan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47419
Availability details for
Quantity
  • Availability
  • Add to Cart