ADVH14DBE254 AChemBlock

(R)-3,3-dimethylcyclohexan-1-amine hydrochloride

  • MDL number MFCD20697046
  • Cas Number : 934765-89-6
  • Molecular Weight : 163.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-3-3-dimethylcyclohexan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44800
Availability details for
Quantity
  • Availability
  • Add to Cart