ADVH14DBE28C AChemBlock

3-aminocyclohexan-1-one

  • MDL number MFCD09029428
  • Cas Number : 149520-74-1
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-aminocyclohexan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48005
Availability details for
Quantity
  • Availability
  • Add to Cart