ADVH14DBE315 AChemBlock

3-methoxybicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD30489919
  • Cas Number : 156329-86-1
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-methoxybicyclo-1-1-1-pentane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47848
Availability details for
Quantity
  • Availability
  • Add to Cart