ADVH14DBE45C AChemBlock

1,1-diethyl 3,3-difluorocyclobutane-1,1-dicarboxylate

  • MDL number MFCD22566196
  • Cas Number : 827032-79-1
  • Molecular Weight : 236.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-1-diethyl-3-3-difluorocyclobutane-1-1-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45132
Availability details for
Quantity
  • Availability
  • Add to Cart