ADVH14DBE4B4 AChemBlock

(2S)-2-methylazetidine hydrochloride

  • MDL number MFCD29920004
  • Cas Number : 935669-67-3
  • Molecular Weight : 107.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2s-2-methylazetidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44797
Availability details for
Quantity
  • Availability
  • Add to Cart