ADVH14DBE4D1 AChemBlock

methyl 3-oxa-7-azabicyclo[3.3.1]nonane-9-carboxylate hydrochloride

  • MDL number MFCD31705796
  • Cas Number : 1536394-89-4
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-oxa-7-azabicyclo-3-3-1-nonane-9-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47926
Availability details for
Quantity
  • Availability
  • Add to Cart