ADVH14DBE548 AChemBlock

1-(3-ethynylazetidin-1-yl)ethan-1-one

  • MDL number MFCD28110671
  • Cas Number : 1824062-27-2
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-3-ethynylazetidin-1-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46893
Availability details for
Quantity
  • Availability
  • Add to Cart