ADVH14DBE54E AChemBlock

(3-fluoro-1-methylazetidin-3-yl)methanol

  • MDL number MFCD31705848
  • Cas Number : 2167753-58-2
  • Molecular Weight : 119.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-1-methylazetidin-3-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46071
Availability details for
Quantity
  • Availability
  • Add to Cart