ADVH14DBE59D AChemBlock

7-fluoro-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid

  • MDL number MFCD12962595
  • Cas Number : 1190315-40-2
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-fluoro-1h-pyrrolo-3-2-c-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49557
Availability details for
Quantity
  • Availability
  • Add to Cart