ADVH14DBE5B0 AChemBlock

(1R,3R)-3-hydroxycyclohexane-1-carboxylic acid

  • MDL number MFCD19228335
  • Cas Number : 1821707-49-6
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-3-hydroxycyclohexane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46948
Availability details for
Quantity
  • Availability
  • Add to Cart