ADVH14DBE610 AChemBlock

(1R,2R)-rel-2-aminocyclohexane-1-carboxylic acid hydrochloride

  • MDL number MFCD05863557
  • Cas Number : 57266-57-6
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-2r-rel-2-aminocyclohexane-1-carboxylic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44580
Availability details for
Quantity
  • Availability
  • Add to Cart