ADVH14DBE679 AChemBlock

Methyl 3-chloro-4-(hydroxymethyl)benzoate

  • MDL number MFCD15527251
  • Cas Number : 1260809-99-1
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-chloro-4-hydroxymethyl-benzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K13431
Availability details for
Quantity
  • Availability
  • Add to Cart