ADVH14DBE67A AChemBlock

2-((benzyloxy)methyl)cyclobutan-1-ol

  • MDL number MFCD31537391
  • Cas Number : 2168976-27-8
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-benzyloxy-methyl-cyclobutan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K13433
Availability details for
Quantity
  • Availability
  • Add to Cart