ADVH14DBE69A AChemBlock

propan-2-yl (2S)-2-{[pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate

  • MDL number MFCD22665799
  • Cas Number : 1256490-52-4
  • Molecular Weight : 453.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/propan-2-yl-2s-2-pentafluorophenoxy-phenoxy-phosphoryl-amino-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P50903
Availability details for
Quantity
  • Availability
  • Add to Cart