ADVH14DBE758 AChemBlock

1,6-naphthyridin-5-ol

  • MDL number MFCD04113798
  • Cas Number : 23616-31-1
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-6-naphthyridin-5-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P51100
Availability details for
Quantity
  • Availability
  • Add to Cart