ADVH14DBE777 AChemBlock

7-(trifluoromethyl)-2-methylquinoline

  • MDL number MFCD11108688
  • Cas Number : 324-32-3
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-trifluoromethyl-2-methylquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P51131
Availability details for
Quantity
  • Availability
  • Add to Cart