ADVH14DBE916 AChemBlock

5-bromo-1-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD17168487
  • Cas Number : 1269293-80-2
  • Molecular Weight : 186.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-1-methyl-1h-pyrazole-4-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P51544
Availability details for
Quantity
  • Availability
  • Add to Cart