ADVH14DBE954 AChemBlock

(S)-2-(((9H-fluoren-9-yl)methoxy)carbonylamino)-3-hydroxy-3-methylbutanoic acid

  • MDL number MFCD04974410
  • Cas Number : 1217603-41-2
  • Molecular Weight : 355.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-9h-fluoren-9-yl-methoxy-carbonylamino-3-hydroxy-3-methylbutanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P51608
Availability details for
Quantity
  • Availability
  • Add to Cart