ADVH1F1C5BBA AChemBlock

tert-butyl N-{4-hydroxybicyclo[2.2.1]heptan-1-yl}carbamate

  • MDL number MFCD31697797
  • Cas Number : 2231676-44-9
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-hydroxybicyclo-2-2-1-heptan-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P50830
Availability details for
Quantity
  • Availability
  • Add to Cart